missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium perchlorate, electrochemical grade

Product Code. 11315068
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11315068

Brand: Thermo Scientific Alfa Aesar 043999.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for use as a polarographic supporting electrolyte | CAS: 1923-70-2 | C16H36ClNO4 | 341.917 g/mol

Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1923-70-2
Molecular Formula C16H36ClNO4
Molecular Weight (g/mol) 341.917
MDL Number MFCD00038722
InChI Key KBLZDCFTQSIIOH-UHFFFAOYSA-M
Synonym tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
PubChem CID 74723
IUPAC Name tetrabutylazanium;perchlorate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 210°C to 217°C
Color White
Linear Formula [CH3(CH2)3]4NClO4
Quantity 250 g
UN Number UN1481
Sensitivity Hygroscopic
Solubility Information Soluble in acetonitrile and ethanol; Very slightly soluble in water.
Formula Weight 341.92
Grade Electrochemical
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium perchlorate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.