missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium iodotetrachloride, 95%

Product Code. 11350114
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350114 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11350114 Supplier Thermo Scientific Alfa Aesar Supplier No. L00574.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15625-59-9 | C16H36Cl4IN | 511.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15625-59-9
Molecular Formula C16H36Cl4IN
Molecular Weight (g/mol) 511.175
MDL Number MFCD00051851
InChI Key HBZJAVINJZUOEH-UHFFFAOYSA-N
Synonym tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide
PubChem CID 73555556
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl

Specifications

Melting Point 135°C to 139°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 511.19
Percent Purity 95%
Chemical Name or Material Tetra-n-butylammonium iodotetrachloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.