missing translation for 'onlineSavingsMsg'
Learn More

Tetra-N-Butylammonium Iodide, 98+%, SLR, Fisher Chemical

Product Code. 10326200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10326200 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10326200 Supplier Fisher Chemical Supplier No. T/0490/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 141°C
CAS 311-28-4
Color White
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula C16H36IN
MDL Number MFCD00011636
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;iodide
Molecular Weight (g/mol) 369.38
PubChem CID 67553
Formula Weight 369.36g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.