missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydroxide, 40% w/w in methanol

Product Code. 11400463
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11400463

Brand: Thermo Scientific Alfa Aesar A12626.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.

Solubility
Miscible with organic solvents.

Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
IUPAC Name tetrabutylazanium;hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.91
Flash Point 18°C (64°F)
Odor Alcohol-like
Refractive Index 1.4
Quantity 2500 mL
UN Number UN2924
Beilstein 3571228
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with organic solvents.
Formula Weight 259.48
Concentration or Composition (by Analyte or Components) 40% w/w in methanol
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.