missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium fluoride trihydrate, 98%

Product Code. 11358016
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358016 50 g 50g
11348016 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358016 Supplier Thermo Scientific Alfa Aesar Supplier No. L13303.18
 Restricted Item
Estimated Shipment: 03-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 87749-50-6 | C16H42FNO3 | 315.514 g/mol

Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 87749-50-6
Molecular Formula C16H42FNO3
Molecular Weight (g/mol) 315.514
MDL Number MFCD00149981
InChI Key VEPTXBCIDSFGBF-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
PubChem CID 11726816
IUPAC Name tetrabutylazanium;fluoride;trihydrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]

Spezifikation

Melting Point 54°C to 60°C
Odor Odorless
Linear Formula [CH3(CH2)3]4NF·3H2O
Quantity 50 g
UN Number UN2811
Beilstein 3761900
Sensitivity Hygroscopic
Solubility Information Soluble in water,terahydrofuran,dimethyl sulfoxide and acetonitrile.
Formula Weight 315.52 (261.47 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium fluoride trihydrate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.