missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium fluoride, 75% w/w aq. soln.

Product Code. 11444797
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444797 500 g 500g
11424797 25 g 25g
11434797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444797 Supplier Thermo Scientific Alfa Aesar Supplier No. B21261.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

Tetrabutylammonium Fluoride is a reactant used for the synthesis of multiple compounds, including conjugated dienoic acid esters, oligoribonucleotides with phosphonate-modified linkages, and triple monoamine reuptake inhibitors, among others. It has proven useful in studying cancer, diabetes, and neurochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabutylammonium Fluoride is a reactant used for the synthesis of multiple compounds, including conjugated dienoic acid esters, oligoribonucleotides with phosphonate-modified linkages, and triple monoamine reuptake inhibitors, among others. It has proven useful in studying cancer, diabetes, and neurochemistry.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
IUPAC Name tetrabutylazanium;fluoride
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Quantity 500 g
UN Number UN2922
Solubility Information Insoluble in water.
Formula Weight 261.47
Concentration or Composition (by Analyte or Components) 75% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.