missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane

Product Code. 11344815
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344815 1 g 1g
11354815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11344815 Supplier Thermo Scientific Alfa Aesar Supplier No. L17891.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 99337-56-1 | C16H38F3N | 301.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Flash Point 13°C (55°F)
Quantity 1 g
UN Number UN2924
IUPAC Name tetrabutylazanium;fluoride;dihydrofluoride
Formula Weight 301.48
Concentration or Composition (by Analyte or Components) 50 to 55% w/w soln. in 1,2-dichloroethane
Chemical Name or Material Tetra-n-butylammonium dihydrogentrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.