missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium difluorotriphenylstannate, 97%, Thermo Scientific™

Product Code. 11339046
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339046 5 g 5g
11329046 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339046 Supplier Thermo Scientific Alfa Aesar Supplier No. L20205.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Can be regarded as a non-hygroscopic anhydrous equivalent of TBAF useful in the preparation of gem-difluorides and the alkylation of enol silyl ethers. For use in the fluorination of carbohydrates.

Solubility
Soluble in most organic solvents.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139353-88-1
Molecular Formula C34H51F2NSn
Molecular Weight (g/mol) 630.496
MDL Number MFCD00192465
InChI Key ODMXVCNGZGLSRS-UHFFFAOYSA-L
Synonym tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetra-n-butylammonium difluorotriphenylstannate, difluorotriphenylstannanuide; tetrabutylammonium, difluoro triphenyl stannanuide; tetrabutylammonium
PubChem CID 2733194
IUPAC Name difluoro(triphenyl)stannanuide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F

Specifications

Melting Point 188°C to 191°C
Quantity 5 g
UN Number UN3146
Beilstein 8668854
Sensitivity Hygroscopic
Solubility Information Soluble in most organic solvents.
Formula Weight 630.5
Percent Purity 97%
Chemical Name or Material Tetra-n-butylammonium difluorotriphenylstannate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.