missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium acetate

Product Code. 11343527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343527 5 g 5g
11353527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343527 Supplier Thermo Scientific Alfa Aesar Supplier No. 019279.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10534-59-5 | C18H39NO2 | 301.52 g/mol

Tetra-n-butylammonium acetate  is used as catalyst, curing agent, and initiator in the polymer industry. It is also used catalyst for the alkynylation of carbonyl compounds with trimethylsilylacetylenes. It is also used as PE/PP clarifying agent, epoxy polymerization initiator, surfactants, emulsifiers, and antistatic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10534-59-5
Molecular Formula C18H39NO2
Molecular Weight (g/mol) 301.52
MDL Number MFCD00043208
InChI Key MCZDHTKJGDCTAE-UHFFFAOYSA-M
Synonym tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
PubChem CID 82707
IUPAC Name tetrabutylazanium;acetate
SMILES CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 114°C to 115°C
Color White
Linear Formula [CH3(CH2)3]4N(O2CCH3)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in acetonitrile (0.1g/mL).
Formula Weight 301.51
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.