missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium acetate, 1.0M aq. soln.

Product Code. 11416506
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416506 25 g 25g
11426506 100 g 100g
11436506 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416506 Supplier Thermo Scientific Alfa Aesar Supplier No. B24773.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10534-59-5 | C18H39NO2 | 301.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10534-59-5
Molecular Formula C18H39NO2
Molecular Weight (g/mol) 301.52
MDL Number MFCD00043208
InChI Key MCZDHTKJGDCTAE-UHFFFAOYSA-M
Synonym tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
PubChem CID 82707
IUPAC Name tetrabutylazanium;acetate
SMILES CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.99
Boiling Point 100°C
Linear Formula [CH3(CH2)3]4N(O2CCH3)
Refractive Index 1.389
Quantity 25 g
Beilstein 3599376
Formula Weight 301.51
Concentration or Composition (by Analyte or Components) 1.0M aq. soln.
Chemical Name or Material Tetra-n-butylammonium acetate
compliance-icons
Product Identifier
  • Tetra-n-butylammonium acetate, 1.0M aqueous solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Tetra-n-butylammoniu

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.