missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butoxysilane, 97%

Product Code. 11352036
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352036 25 g 25g
11362036 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352036 Supplier Thermo Scientific Alfa Aesar Supplier No. L03141.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4766-57-8 | C16H36O4Si | 320.545 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4766-57-8
Molecular Formula C16H36O4Si
Molecular Weight (g/mol) 320.545
MDL Number MFCD00009432
InChI Key UQMOLLPKNHFRAC-UHFFFAOYSA-N
Synonym tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
PubChem CID 78500
IUPAC Name tetrabutyl silicate
SMILES CCCCO[Si](OCCCC)(OCCCC)OCCCC

Specifications

Boiling Point 277°C
Melting Point <-80°C
Quantity 25 g
Beilstein 1709274
Flash Point 135°C (275°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 320.55
Percent Purity 97%
Odor Characteristic
Refractive Index 1.413
Sensitivity Moisture sensitive
Density 0.899
Chemical Name or Material Tetra-n-butoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.