missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetra Core Set and Novex™ Tris-Glycine 4-20% Gels Combos

Product Code. 30484842
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30484842

Brand: Invitrogen™ XP0420TS10W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Tetra Electrophoresis Core Set and Novex Tris-Glycine 4-20% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of Novex Tris-Glycine gels. Achieve broad-range protein separation with Laemmli-like chemistry in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank, running up to four gels per run.

The Invitrogen Tetra Electrophoresis Core Set and Novex Tris-Glycine 4-20% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of Novex Tris-Glycine gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. The intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of Novex Tris-Glycine 4-20% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel in the core. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About Novex Tris-Glycine gels, WedgeWell format

Novex Tris-Glycine mini gels are polyacrylamide gels based on traditional Laemmli protein electrophoresis, allowing the use of Laemmli sample and running buffers. They enable consistent performance and separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using for Novex Tris-Glycine Native Running Buffer (Cat. No. LC2672) or Novex Tris-Glycine Transfer Buffer (Cat. No. LC3675) for traditional wet transfer using the XCell II Blot Module (Cat. No. EI9051). Alternatively, rapid semi-dry transfer can be performed using the Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (Cat. No. IB31001).

TRUSTED_SUSTAINABILITY

Specifications

Adapters Tetra Electrophoresis Cores
Content And Storage Store at 2°C to 8°C. Do not freeze.
Buffer Novex Tris-Glycine SDS Running Buffer, Novex Tris-Glycine Native Running Buffer
For Use With (Equipment) Bio-Rad Mini Protean Tanks, Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 4 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
Product Type Electrophoresis Cores and Precast Protein Gels
Sample Loading Volume Up to 60 μL
Separation of Protein
Separation Range 20 to 200 kDa
Separation Type Denaturing, Native
Wells 10-well
Mode of Separation Molecular Weight
Product Line Novex, WedgeWell
Quantity Tetra Core Set, 3 boxes of gels (30 gels)
Shipping Condition Wet Ice
Length (Metric) 8 cm
Width (Metric) 8 cm
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.