missing translation for 'onlineSavingsMsg'
Learn More

TetR Antibody, Allophycocyanin, Novus Biologicals™

Product Code. 18409300 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409300 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409300 Supplier Novus Biologicals Supplier No. NBP178424APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TetR Polyclonal antibody specifically detects TetR in Nonspecies specific samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TetR
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of Escherichia coli TetR protein (between residues 1-50) [UniProt P04483]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4924774
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.