missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetanus Toxin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-62945JF549
206.55 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for ELISA
Specifications
| Tetanus Toxin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Janelia Fluor 549 | |
| Tentoxylysin | |
| Synthetic peptide corresponding to the amino acids 1300 - 1314 of the native molecule conjugated to Keyhole limpet Haemocyanin | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction