missing translation for 'onlineSavingsMsg'
Learn More

TETA, TRC

Product Code. 30413223
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413223 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413223 Supplier Toronto Research Chemicals Supplier No. TRCT20453510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TETA
CAS 314041-07-1
Molecular Formula C18H32N4O8 • 4 HCl • x H2O
Quantity 10 mg
Synonym 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic acid tetrahydrochloride','monohydrate
InChI Formula InChI=1S/C18H32N4O8.4ClH.H2O/c23-15(24)11-19-3-1-4-20(12-16(25)26)8-10-22(14-18(29)30)6-2-5-21(9-7-19)13-17(27)28;;;;;/h1-14H2,(H,23,24)(H,25,26)(H,27,28)(H,29,30);4*1H;1H2
SMILES O=C(CN1CCN(CCCN(CCN(CCC1)CC(O)=O)CC(O)=O)CC(O)=O)O.Cl.O.Cl.Cl.Cl
IUPAC Name 2,2',2'',2'''-(1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl)tetraacetic acid tetrahydrochloride hydrate
Molecular Weight (g/mol) 596.33
Formula Weight 594.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.