missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33311-100ul
This item is not returnable.
View return policy
Description
TET3 Monoclonal antibody specifically detects TET3 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| TET3 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of mouse TET3 (Q8BG87).,, Sequence:, MSQFQVPLAVQPDLSGLYDFPQGQVMVGGFQGPGLPMAGSETQLRGGGDGRKKRKRCGTCDPCRRLENCGSCTSCTNRRTHQICKLRKCEVLKKKAGLLKEVEINAREGTGPWAQGATVKTGSELSPVDGPVPGQMDSGPVYHGDSRQLSTSGAPVNGAREPAGPGLLGAAGPWRVDQKPDWEAASGPTHAARLEDAHDLVAFSAVAEAVSSYGALSTRLYETFNREMSREAGSNGRGPRPESCSEGSEDLDTLQTALALARHGMKPPNCTCDGPECPDFLEWLE | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 200424 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction