missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82066JF549
This item is not returnable.
View return policy
Description
TET3 Polyclonal antibody specifically detects TET3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TET3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
| TET3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human TET3. The immunogen is located within amino acids 1520 - 1570 of TET3. Amino Acid Squence: ARRELHATTPLKKPNR | |
| 0.1 mL | |
| Epigenetics | |
| 200424 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction