missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 54790 |
| Species | Human |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-27465. For further blocking peptide related protocol, click here . |
| Gene Alias | FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 |
| Gene Symbol | TET2 |
| Quantity | 0.1 mg |
| Storage Buffer | Lyophilized from 5 pmol/μL in 5% (v/v) acetonitrile/H2O. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?