missing translation for 'onlineSavingsMsg'
Learn More

TET1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18419954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419954 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419954 Supplier Novus Biologicals Supplier No. NBP178966JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TET1 Polyclonal antibody specifically detects TET1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot (Negative), ICC/IF (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TET1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot (Negative), ICC/IF (Negative)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800). [UniProt Q8NFU7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 80312
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.