missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Undecanoate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Testosterone Undecanoate |
| CAS | 5949-44-0 |
| Molecular Formula | C30 H48 O3 |
| Quantity | 10 mg |
| Synonym | Androst-4-en-3-one','17-[(1-oxoundecyl)oxy]-','(17β)-, Testosterone','undecanoate (6CI,7CI,8CI), Undecanoic acid','ester with testosterone (8CI), Andriol, Andriol Testocaps, Nebido, Org 538, Testocaps, Testosterone undecylate, Undestor |
| InChI Formula | InChI=1S/C30H48O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h21,24-27H,4-20H2,1-3H3/t24-,25-,26-,27-,29-,30-/m0/s1 |
| SMILES | CCCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] undecanoate |
| Molecular Weight (g/mol) | 456.7 |
| Formula Weight | 456.3603 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction