missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Testosterone Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175023
This item is not returnable.
View return policy
Description
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL 17Beta testosterone produces 65% inhibition in a competitive ELISA employing 17βTestosterone polyclonal antibody.
Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
Specifications
| Testosterone | |
| Polyclonal | |
| Unconjugated | |
| Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron | |
| 17b-Testosterone(7)-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 7.56 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction