missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Enanthate, TRC

Product Code. 30428486
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428486 Supplier Toronto Research Chemicals Supplier No. TRCT155080100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone enanthate
CAS 315-37-7
Molecular Formula C26 H40 O3
Quantity 100 mg
Synonym Testosterone Enantate, Testosterone Isocaproate Imp. F (EP), Androst-4-en-3-one','17-[(1-oxoheptyl)oxy]-','(17β)-, Testosterone','heptanoate (6CI,8CI), (17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, Heptanoic acid','ester with testosterone (6CI), 17β-Enanthoxyandrost-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one enanthate, 4-Androsten-3-one 17β-enanthate, Andro LA 200, Androtardyl, Delatestryl, Enarmone depot, Everone, Jenasteron, NSC 17591, Primoteston-Depot, Reposo TMD, Testenate, Testinon, Testo-Enant, Testosterone 17-enanthate, Testosterone enanthate, Testosterone heptylate, Testosterone oenanthate
InChI Formula InChI=1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1
SMILES CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate
Molecular Weight (g/mol) 400.59
Formula Weight 400.2977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.