missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Cypionate-d3, TRC

Product Code. 30416607
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416607 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416607 Supplier Toronto Research Chemicals Supplier No. TRCT15510210MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Cypionate-d3
CAS 876054-56-7
Molecular Formula C27 D3 H37 O3
Quantity 10 mg
Synonym Androst-4-en-3-one-16,16,17-d3','17-(3-cyclopentyl-1-oxopropoxy)-','(17β)- (9CI), (17β)-17-(3-Cyclopentyl-1-oxopropoxy)androst-4-en-3-one-16,16,17-d3, Testosterone-16,16,17-d3 17-cyclopentanepropionate, 17β-Hydroxyandrost-4-en-3-one-16,16,17-d3 cyclopentylpropionate, Testosterone Cypionate-d3, Testosterone Cypionate-16,17,17-D3
InChI Formula InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1/i11D2,24D
SMILES [2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Molecular Weight (g/mol) 415.623
Formula Weight 415.317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.