missing translation for 'onlineSavingsMsg'
Learn More

Testosterone cypionate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30322941
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322941 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322941 Supplier Dr. Ehrenstorfer Supplier No. DREA17322530AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Cypionate
CAS 58-20-8
Analyte or Component Names Testosterone Cypionate
Shipping Condition Room Temperature
Molecular Formula C27 H40 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Testosterone, cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid, ester with testosterone (8CI), DepAndro 100, DepAndro 200, Depo, Depo-testosterone, Deposteron, Depotest, Depovirin, NSC 9157, Pertestis, Testex Leo, Testodrin prolongatum, Testosterone cyclopentylpropionate, Testosterone cypionate, Testosterone, 3-cyclopentanepropionate, Virilon, Androst-4-en-3-one, 17-(3-cyclopentyl-1-oxopropoxy)-, (17beta)-, 17beta-Hydroxyandrost-4-en-3-one cyclopentanepropionate, 17beta-Hydroxyandrost-4-en-3-one cyclopentylpropionate, Testosterone 17beta-cyclopentanepropionate, Testosterone 17beta-cyclopentylpropionate, Testosterone 17beta-cypionate
InChI Formula InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 412.6
Quantity 1 mL
Formula Weight 412.2977 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.