missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Antibody (4E1G2) - Azide and BSA Free, Novus Biologicals™

Product Code. 18361632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361632 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361632 Supplier Novus Biologicals Supplier No. NBP311603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen Testosterone
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Monoclonal
Clone 4E1G2
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Radioimmunoassay
Formulation 10 mM PBS
Gene Alias 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone
Host Species Mouse
Immunogen Testosterone 3 CMO conjugated to BSA
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.