missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Antibody (3T16), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50332CL1
This item is not returnable.
View return policy
Description
Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA,Radioimmune Assays
Specifications
| Testosterone | |
| Monoclonal | |
| CoraFluor 1 | |
| 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone | |
| Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Radioimmune Assays (RIA) | |
| 3T16 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction