missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Acetate, LoGiCal

Product Code. 30461340
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461340 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461340 Supplier LoGiCal Supplier No. LGCFOR0323.05
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Acetate
CAS 1045-69-8
Analyte or Component Names Testosterone Acetate
Shipping Condition Room Temperature
Molecular Formula C21 H30 O3
Certifications/Compliance ISO 9001
Synonym Testosterone Imp. E (EP), Testosterone Isocaproate Imp. B (EP), Testosterone Propionate Imp. A (EP), Testosterone Acetate, 3-Oxoandrost-4-en-17beta-yl Acetate, (17beta)-17-(Acetyloxy)androst-4-en-3-one, Testosterone acetate, 17-O-Acetyltestosterone, 17beta-Acetoxy-4-androsten-3-one, 17beta-Acetoxy-Delta4-androsten-3-one, 17beta-Acetoxyandrost-4-ene-3-one, 17beta-Acetoxytestosterone, 17beta-Hydroxyandrost-4-en-3-one acetate, 17beta-Hydroxyandrost-4-en-3-one-17beta-acetate, 3-Oxoandrost-4-en-17beta-yl acetate, 4-Androsten-17beta-ol-3-one 17-acetate, Aceto-sterandryl, Aceto-testoviron, Amolisin, Androst-4-en-17beta-ol-3-one acetate, Androtest A, Deposteron, Farmatest, NSC 523836, Perandrone A, SKF 5647, Testosterone 17-acetate, Testosterone Impurity E, Testosterone Isocaproate Impurity B, Testosterone Propionate Impurity A
InChI Formula InChI=1S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h12,16-19H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21-/m0/s1
Solution Type Neat
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 330.46
Quantity 10 mg
Formula Weight 330.2195 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.