missing translation for 'onlineSavingsMsg'
Learn More

Testosterone 17-Phenylpropionate, TRC

Product Code. 30423049
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423049 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423049 Supplier Toronto Research Chemicals Supplier No. TRCT155150250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Phenylpropionate
CAS 1255-49-8
Molecular Formula C28 H36 O3
Quantity 250 mg
Synonym Androst-4-en-3-one','17-(1-oxo-3-phenylpropoxy)-','(17β)-, Testosterone','hydrocinnamate (6CI,7CI,8CI), (17β)-17-(1-Oxo-3-phenylpropoxy)androst-4-en-3-one, Hydrocinnamic acid','ester with testosterone (8CI), 3-Oxoandrost-4-en-17β-yl β-phenylpropionate, Androject, Hydrocinnamic acid','testosterone ester, NSC 26643, Retandrol, Testosterone 17-phenylpropionate, Testosterone phenpropionate, Testosterone phenylpropanoate, Testosterone phenylpropionate
InChI Formula InChI=1S/C28H36O3/c1-27-16-14-21(29)18-20(27)9-10-22-23-11-12-25(28(23,2)17-15-24(22)27)31-26(30)13-8-19-6-4-3-5-7-19/h3-7,18,22-25H,8-17H2,1-2H3/t22-,23-,24-,25-,27-,28-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCc5ccccc5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate
Molecular Weight (g/mol) 420.58
Formula Weight 420.2664
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.