missing translation for 'onlineSavingsMsg'
Learn More

Testosterone 17-O-Acetate, TRC

Product Code. 30415956
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415956 Supplier Toronto Research Chemicals Supplier No. TRCT155025100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Acetate
CAS 1045-69-8
Molecular Formula C21 H30 O3
Quantity 100 mg
Synonym Testosterone Acetate, 3-Oxoandrost-4-en-17beta-yl Acetate, Testosterone Propionate Imp. A (EP), Testosterone Isocaproate Imp. B (EP), Testosterone Imp. E (EP), (17β)-17-(Acetyloxy)androst-4-en-3-one, Testosterone acetate, 17-O-Acetyltestosterone, 17β-Acetoxy-4-androsten-3-one, 17β-Acetoxy-Δ4-androsten-3-one, 17β-Acetoxyandrost-4-ene-3-one, 17β-Acetoxytestosterone, 17β-Hydroxyandrost-4-en-3-one acetate, 17β-Hydroxyandrost-4-en-3-one-17β-acetate, 3-Oxoandrost-4-en-17β-yl acetate, 4-Androsten-17β-ol-3-one 17-acetate, Aceto-sterandryl, Aceto-testoviron, Amolisin, Androst-4-en-17β-ol-3-one acetate, Androtest A, Deposteron, Farmatest, NSC 523836, Perandrone A, SKF 5647, Testosterone 17-acetate
InChI Formula InChI=1S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h12,16-19H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21-/m0/s1
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 330.46
Formula Weight 330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.