missing translation for 'onlineSavingsMsg'
Learn More

Testosterone 17-Isobutyrate, TRC

Product Code. 30422522
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30422522

Brand: Toronto Research Chemicals TRCT15503510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate)
CAS 1169-49-9
Molecular Formula C23 H34 O3
Quantity 10 mg
Synonym 3-Oxoandrost-4-en-17beta-yl 2-Methylpropanoate, Testosterone Propionate Imp. B (EP), Testosterone Isocaproate Imp. D (EP), Testosterone isobutyrate, Androst-4-en-3-one','17-(2-methyl-1-oxopropoxy)-','(17β)-, Testosterone','isobutyrate (6CI,7CI,8CI), (17β)-17-(2-Methyl-1-oxopropoxy)androst-4-en-3-one, Isobutyric acid','ester with testosterone (8CI), 17β-(Isobutyroxy)androst-4-en-3-one, 3-Oxoandrost-4-en-17β-yl isobutyrate, Agovirin-Depot, Perandren M, Testocryst, Testosterone 17-isobutyrate, Testosterone 2-methylpropanoate, Testosterone 2-methylpropionate
InChI Formula InChI=1S/C23H34O3/c1-14(2)21(25)26-20-8-7-18-17-6-5-15-13-16(24)9-11-22(15,3)19(17)10-12-23(18,20)4/h13-14,17-20H,5-12H2,1-4H3/t17-,18-,19-,20-,22-,23-/m0/s1
SMILES CC(C)C(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 2-methylpropanoate
Molecular Weight (g/mol) 358.51
Formula Weight 358.2508
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.