missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testisin/Prss21 Antibody (2E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010942-M01
This item is not returnable.
View return policy
Description
Testisin/Prss21 Monoclonal antibody specifically detects Testisin/Prss21 in Human samples. It is validated for ELISA, Immunoprecipitation
Specifications
| Testisin/Prss21 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 3.4.21, Eosinophil serine protease 1, ESP1EC 3.4.21.-, ESP-1serine protease from eosinophils, protease, serine, 21 (testisin), Serine protease 21, TEST1testisin, TESTISIN | |
| PRSS21 (NP_006790, 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VSNIYLSPRYLGNSPYDIALVKLSAPVTYTKHIQPICLQASTFEFENRTDCWVTGWGYIKEDEALPSPHTLQEVQVAIINNSMCNHLFLKYSFRKDIFGD | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation | |
| 2E10 | |
| ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| NP_006790 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 10942 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction