missing translation for 'onlineSavingsMsg'
Learn More

Testin Antibody (1G11-B7), Novus Biologicals™

Product Code. 18319809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319809 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319809 Supplier Novus Biologicals Supplier No. H00026136M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Testin Monoclonal antibody specifically detects Testin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Testin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G11-B7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01451
Gene Alias DKFZp586B2022, MGC1146, TESS, TESS-2, testin, testis derived transcript (3 LIM domains)
Host Species Mouse
Immunogen TES (AAH01451, 1 a.a. ~ 421 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDLENKVKKMGLGHEQGFGAPCLKCKEKCEGFELHFWRKICRNCKCGQEEHDVLLSNEEDRKVGKLFEDTKYTTLIAKLKSDGIPMYKRNVMILTNPVAAKKNVSINTVTYEWAPPVQNQALARQYMQMLPKEKQPVAGSEGAQYRKKQLAKQLPAHDQDPSKCHELSPREVKEMEQFVKKYKSEALGVGDVKLPCEMDAQGPKQMNIPGGDRSTPAAVGAMEDKSAEHKRTQYSCYCCKLSMKEGDPAIYAERAGYDKLWHPACFVCSTCHELLVDMIYFWKNEKLYCGRHYCDSEKPRCAGCDELIFSNEYTQAENQNWHLKHFCCFDCDSILAGEIYVMVNDKPVCKPCYVKNHAVVCQGCHNAIDPEVQRVTYNNFSWHASTECFLCSCCSKCLIGQKFMPVEGMVFCSVECKKRMS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26136
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.