missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ ECO 8 Sample Test Tube

Product Code. 12393307 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
ECO 8 Thermoreactor
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12393307 ECO 8 Thermoreactor Each
1 options
This item is not returnable. View return policy

Product Code. 12393307

Brand: Velp Scientifica™ CA0091666

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Velp Scientifica™ ECO 8 Sample Test Tube for sample decomposition.

  • Sampling test tube
  • For use with ECO 8 Thermoreactor

Specifications

Format Test Tube
For Use With (Equipment) ECO 8 Thermoreactor
Includes NS 19/26 cone, Glass cap
Quantity 1 Each
Diameter (Metric) Outer 22 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.