Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 30181
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11436358
  • £314.08 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Specifications
Test Tube Rack
6 x 6
10 to 13 mm Tubes
MaxQ™ Shaker Platform
Red; 10 to 13 mm; 6 x 6 array
Red
36