Learn More
Memmert™ Test Tube Racks with 40 Holes
Test tube rack with 40 holes and 2-row rack
Brand:  Memmert™ WTB15B48825
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17190315
  • £54.25 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • Test tube rack
  • 40 holes 2-row rack
Specifications
Specifications
40 holes, 2-row rack
Test Tube Racks
12 mm
WTB15
Longitudinals for sizes 15 and 24