Learn More
Memmert™ Test Tube Racks with 20 Holes
Shop All Memmert Products
Click to view available options
Features:
Longitudinals for sizes 15 and 24
Transverses for sizes 15 and 24
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB6
Unit Size:
Each
Description
- Test tube rack
- 20 holes 2-row rack
- Diameter is 21 mm
Specifications
Specifications
| Description | 20 holes, 2-row rack |
| For Use With (Equipment) | WTB6 |
| Type | Test Tube Rack |
| Features | Transverses for sizes 15 and 24 |
| Diameter (Metric) | 21 mm |