Learn More
Memmert™ Test Tube Racks with 12 Holes
Product Code. 17100425 Shop All Memmert Products
Click to view available options
Features:
Longitudinals for sizes 15 and 24
Transverses for sizes 15 and 24
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB6
Unit Size:
Each
Product Code. 17100425
Brand: Memmert™ WTB6B48832
Currently Unavailable: We have no information when this product will be available.
View alternative products
Test tube rack with 12 holes and 2-row rack
  • Test tube rack
  • 12 holes 2-row rack
  • Diameter is 32 mm
Specifications
Description 12 holes, 2-row rack
For Use With (Equipment) WTB6
Type Test Tube Rack
Features Transverses for sizes 15 and 24
Diameter (Metric) 32 mm