Learn More
Julabo™ Test Tube Rack
Click to view available options
No. of Positions:
21
30
42
60
90
Unit Size:
Each
Specifications
Specifications
| Description | Test tube racks for water baths |
| Temperature Range | Up to +100°C |
| Material | High grade plastic |
| No. of Positions | 90 |
| Size | 75 x 12/13 mm dia. |