missing translation for 'onlineSavingsMsg'
Learn More

JULABO™ Test Tube Rack

Product Code. 15410642
Click to view available options
Capacity (Metric):
42 tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15410642

Brand: JULABO™ 9970322

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Test tube rack for 42 tubes

  • 40 x 10/11 mm dia. tubes
  • Made of stainless steel, up to +150 °C

Specifications

Description Max temperature 150°C
Capacity (Metric) 42 tubes
For Use With (Equipment) CORIO™ C-B5, CORIO™ C-BT5, CORIO™ CD-200F, CORIO™ CD-201F, CORIO™ CD-300F, CORIO™ CD-B5, CORIO™ CD-BT5, CORIO™ CP-200F, CORIO™ CP-201F, CORIO™ CP-300F, CORIO™ CP-BC4, CORIO™ CP-BC6, DYNEO™ DD-200F, DYNEO™ DD-201F, DYNEO™ DD-300F
Material Stainless steel
No. of Positions 42
Diameter (Metric) 10/11 mm dia.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.