Learn More
Memmert™ Test Tube Rack
Product Code. 17592265 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
WTB35 Water Baths
WTB50 Water Baths
Holds:
100 Test Tubes (4 Rows)
30 Test Tubes (2 Rows)
44 Test Tubes (2 Rows)
50 Test Tubes (2 Rows)
58 Test Tubes (2 Rows)
66 Test Tubes (2 Rows)
74 Test Tubes (2 Rows)
75 Test Tubes (3 Rows)
Diameter (Metric):
10 mm
12 mm
14.5 mm
18 mm
21 mm
32 mm
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Holds Diameter (Metric) Unit Size
17592265 WTB50 Water Baths 30 Test Tubes (2 Rows) 32 mm Each
17512265 WTB35 Water Baths 30 Test Tubes (2 Rows) 32 mm Each
17502265 WTB35 Water Baths 44 Test Tubes (2 Rows) 21 mm Each
17592255 WTB35 Water Baths 50 Test Tubes (2 Rows) 18 mm Each
17582255 WTB35 Water Baths 58 Test Tubes (2 Rows) 14.5 mm Each
17572255 WTB35 Water Baths 66 Test Tubes (2 Rows) 12 mm Each
17562255 WTB35 Water Baths 74 Test Tubes (2 Rows) 10 mm Each
17522265 WTB35 Water Baths 75 Test Tubes (3 Rows) 18 mm Each
17532265 WTB35 Water Baths 100 Test Tubes (4 Rows) 18 mm Each
17582265 WTB50 Water Baths 44 Test Tubes (2 Rows) 21 mm Each
17572265 WTB50 Water Baths 50 Test Tubes (2 Rows) 18 mm Each
17562265 WTB50 Water Baths 58 Test Tubes (2 Rows) 14.5 mm Each
17552265 WTB50 Water Baths 66 Test Tubes (2 Rows) 12 mm Each
17542265 WTB50 Water Baths 74 Test Tubes (2 Rows) 10 mm Each
17502275 WTB50 Water Baths 75 Test Tubes (3 Rows) 18 mm Each
17512275 WTB50 Water Baths 100 Test Tubes (4 Rows) 18 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 17592265 Supplier Memmert™ Supplier No. WTB50B50661
  • ONLINE EXCLUSIVE PRICE
    £86.15
    £84.64 / Pack of 1
    Save £1.51
    1% Off
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Test Tube Rack designed for use with a Memmert™ Water Bath.
  • Water Bath Accessory
Specifications
For Use With (Equipment) WTB50 Water Baths
Type Test Tube Rack
Holds 30 Test Tubes (2 Rows)
Material Stainless Steel
Diameter (Metric) 32 mm