Learn More
Memmert™ Test Tube Rack
Product Code. 17522265 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB35 Water Baths
WTB50 Water Baths
Holds:
100 Test Tubes (4 Rows)
30 Test Tubes (2 Rows)
44 Test Tubes (2 Rows)
50 Test Tubes (2 Rows)
58 Test Tubes (2 Rows)
66 Test Tubes (2 Rows)
74 Test Tubes (2 Rows)
75 Test Tubes (3 Rows)
Diameter (Metric):
10 mm
12 mm
14.5 mm
18 mm
21 mm
32 mm
Unit Size:
Each
Product Code. 17522265
Brand: Memmert™ WTB35B50662
Only null left
Add to basket
Add to basket
Test Tube Rack designed for use with a Memmert™ Water Bath.
  • Water Bath Accessory
Specifications
For Use With (Equipment) WTB35 Water Baths
Type Test Tube Rack
Holds 75 Test Tubes (3 Rows)
Material Stainless Steel
Diameter (Metric) 18 mm