missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scientific Industries Test Tube Rack
Click to view available options
Packaging Quantity:
1 Ea.
2 Tube Racks
Unit Size:
Pack of 12
Description
- Holds up to 12 tubes per rack for the Multi Vortex-Genie
- Accommodates tubes up to 29 mm diameter of any material type
- Sold in 1- or 2-pack
Specifications
Specifications
| Holds | 12 Tubes per rack |
| Tube Size | ≤29 mm OD |
| Product Type | Tube Rack |
| Color | Clear |
| For Use With (Equipment) | Multi Vortex-Genie |
| Packaging Quantity | 2 Tube Racks |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction