missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Test Tube Rack

Product Code. 11311494
Change view
Click to view available options
Packaging Quantity:
1 Ea.
2 Tube Racks
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
11311494 1 Ea. Pack of 12
11321494 2 Tube Racks Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311494 Supplier Scientific Industries Supplier No. SIM010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Test Tube Rack holds up to 12 tubes for the Multi Vortex-Genie. Accommodates tubes of any material type, up to 29 mm in diameter.

  • Holds up to 12 tubes per rack for the Multi Vortex-Genie
  • Accommodates tubes up to 29 mm diameter of any material type
  • Sold in 1- or 2-pack
TRUSTED_SUSTAINABILITY

Specifications

Holds 12 Tubes per rack
Tube Size ≤29 mm OD
Product Type Tube Rack
Color Clear
For Use With (Equipment) Multi Vortex-Genie
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.