Learn More
Elma™ Test Tube Holders
Click to view available options
Model:
100/120
150/300
30/80
Unit Size:
Each
Description
- Diverse Elma holders ensure that tubes can be securely positioned and arranged in the Elma Basket
- Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
- Allows items to be individually positioned
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
- Holders are available for various applications
- Special solutions are available (e.g. for conical flasks)
Specifications
Specifications
| Type | Test Tube Holder |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Material | Stainless Steel |
| Model | 150/300 |