missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Test Tube Attachment for IKA Vortex Shaker

Product Code. 10367702 Shop All IKA Products
Click to view available options
Holds:
12 x 12 mm Reagent Glasses
18 x 10 mm Reagent Glasses
8 x 16 mm Reagent Glasses
8 x 20 mm Reagent Glasses
Product Type:
Test Tube Adapter Attachment
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10367702

Brand: IKA™ 0003344000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Attachment with customized array for holding reagent glasses on a vortexed.

  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
TRUSTED_SUSTAINABILITY

Specifications

Holds 12 x 12 mm Reagent Glasses
Tube Size 12 mm
Product Type Test Tube Adapter Attachment
Description Test tube adapter attachment
Model Model VG 3.33
For Use With (Equipment) VG 3.3 Universal Attachment, Vortexer,
No. of Positions 12
Requires VG 3.3 Universal Attachment
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.