Learn More
Hanna Instruments™ Total Chlorine Reagents
Click to view available options
Range:
0 to .35 mg/L
0 to 1 mg/L
0 to 15 mg/L
0 to 2.5 mg/L
0 to 5 mg/L
Unit Size:
100g
Pack of 100
Description
The HI-93721-01 are reagents for the colorimetric determination of iron. There are enough reagents for 100 tests and are for use with Hanna’s compatible benchtop and portable photometers. These high quality reagents are clearly marked with the lot number and expiry date on each packet for traceability.
Specifications
Specifications
| Quantity | 100 Pack |
| Range | 0 to 5 mg/L |
Safety and Handling
Product Identifier
- Iron HR Reagent
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sodium metabisulfite, Sodium dithionite,1,10-Phenanthroline