Learn More
Palintest™ Test Chlorine DPD No 1 + No 3
Product Code. 13101541
Click to view available options
Quantity:
250/Pk.
50/Pk.
Unit Size:
250 tests
50 tests
Product Code. 13101541
Brand: Palintest™ CD011/1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With interchangeable colour discs
The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Packaging 250/Pk.
Quantity 250/Pk.