missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TESPA1 Polyclonal antibody specifically detects TESPA1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | TESPA1 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hypothetical protein LOC9840, KIAA0748 |
| Host Species | Rabbit |
| Immunogen | TESPA1 antibody was raised against an 18 amino acid peptide near the amino terminus of human TESPA1. The immunogen is located within amino acids 20 - 70 of TESPA1. Amino Acid Squence: LQDVPDPEPSSLDDVFQE |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?