Learn More
Invitrogen™ TESK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110474
Description
Immunogen sequence: FVEIGKTLEE ILSRLQEEEQ ERDRKLQPTA RGLLEKAPGV KRLSSLDDKI PHKSPCPRRT IWLSRSQSDI FSRKPPRTVS VLDPYYRPRD GAARTPKVNP FSARQDLMGG KIKFFDLPSK SVISLVFDLD APGPGTMPLA DWQEPLAPPI RRWRSLPGSP EFLHQEACPF VGREESLSDG PPPRLSSLKY RVKEIPPFRA SALPAAQAHE AMDCSILQEE NGFGSRPQGT SPCPAGASEE MEVEERPAGS TPATFSTSGI GLQTQGKQDG.
TESK2 or testis-specific kinase 2 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. The kinase domain of TESK2 is structurally similar to the kinase domain of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). TESK2 is predominantly expressed in testis and prostate and plays an important role in spermatogenesis. TESK2 phosphorylates cofilin specifically at Ser-3 and induces formation of actin stress fibers and focal adhesions.
Specifications
| TESK2 | |
| Polyclonal | |
| Unconjugated | |
| TESK2 | |
| dual specificity testis-specific protein kinase 2; TESK2; Testicular protein kinase 2; testis-specific kinase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10420, 230661 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.155 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VCT9, Q96S53 | |
| TESK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 302-571 of human TESK2 (NP_009101.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.