missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TESK1 Polyclonal Antibody

TESK1 Polyclonal Antibody
Brand: Invitrogen PA5101564
This item is not returnable.
View return policy
Description
TESK1 Polyclonal Antibody for Western Blot
Specifications
| TESK1 | |
| Polyclonal | |
| Unconjugated | |
| TESK1 | |
| AI326901; dual specificity testis-specific protein kinase 1; Tesk1; Testicular protein kinase 1; testis specific protein kinase 1; testis-specific kinase 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21754, 29460, 7016 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70146, Q15569, Q63572 | |
| TESK1 | |
| A synthesized peptide derived from human TESK1(Accession Q15569), corresponding to amino acid residues R185-D235. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction